Assay Details
| Gene Symbol: STEAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077847.1 | NP_001071315.1 | 2 - 3 | 729 | 96 | |
| GenBank mRNA | BC126751.1 | - | 2 - 3 | 729 | 96 | |
More Information
Set Membership: |
|

STEAP2
Genomic Map