Assay Details
Gene Symbol: GRSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077971.1 | NP_001071439.1 | 6 - 7 | 1196 | 70 | |
GenBank mRNA | BC126703.1 | - | 6 - 7 | 1196 | 70 | |
BC149233.1 | - | 6 - 7 | 843 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
