Assay Details
| Gene Symbol: PRIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079594.2 | NP_001073062.1 | 6 - 7 | 661 | 65 | |
| GenBank mRNA | BC126657.1 | - | 6 - 7 | 662 | 65 | |
More Information
Set Membership: |
|

PRIM2
Genomic Map