Assay Details
| Gene Symbol: TMEM104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079596.1 | NP_001073064.1 | 4 - 5 | 353 | 57 | |
| GenBank mRNA | BC126661.1 | - | 4 - 5 | 353 | 57 | |
More Information
Set Membership: |
|

TMEM104
Genomic Map