Assay Details
| Gene Symbol: SMG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080286.1 | NP_001073755.1 | 1 - 2 | 1808 | 124 | |
| GenBank mRNA | BC126609.1 | - | 1 - 2 | 1808 | 124 | |
More Information
Set Membership: |
|

SMG8
Genomic Map