Assay Details
| Gene Symbol: SPRY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081512.1 | NP_001074981.1 | 1 - 2 | 169 | 67 | |
| GenBank mRNA | BC133401.1 | - | 1 - 2 | 169 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC133401.1, NM_001081512.1 transcripts.
Set Membership: |
|

SPRY4
Genomic Map