Assay Details
| Gene Symbol: GATAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081623.2 | NP_001075092.1 | 4 - 5 | 768 | 127 | |
| GenBank mRNA | BC133475.1 | - | 4 - 5 | 769 | 127 | |
More Information
Set Membership: |
|

GATAD1
Genomic Map