Assay Details
| Gene Symbol: LSR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083394.1 | NP_001076863.1 | 2 - 3 | 467 | 53 | |
| GenBank mRNA | BC123896.1 | - | 2 - 3 | 467 | 53 | |
More Information
Set Membership: |
|

LSR
Genomic Map