Assay Details
| Gene Symbol: SNAPIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083410.2 | NP_001076879.1 | 1 - 2 | 163 | 64 | |
| GenBank mRNA | BC114837.1 | - | 1 - 2 | 165 | 64 | |
More Information
Set Membership: |
|

SNAPIN
Genomic Map