Assay Details
| Gene Symbol: CYP27A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083413.2 | NP_001076882.1 | 2 - 3 | 474 | 62 | |
| GenBank mRNA | BC123878.1 | - | 2 - 3 | 475 | 62 | |
More Information
Set Membership: |
|

CYP27A1
Genomic Map