Assay Details
| Gene Symbol: HIF1AN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083443.2 | NP_001076912.1 | 4 - 5 | 740 | 75 | |
| GenBank mRNA | BC133607.1 | - | 4 - 5 | 741 | 75 | |
More Information
Set Membership: |
|

HIF1AN
Genomic Map