Assay Details
| Gene Symbol: TFF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083521.1 | NP_001076990.1 | 3 - 4 | 395 | 64 | |
| GenBank mRNA | BC114896.1 | - | 3 - 4 | 395 | 64 | |
More Information
Set Membership: |
|

TFF2
Genomic Map