Assay Details
| Gene Symbol: MEDAG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083660.1 | NP_001077129.1 | 1 - 2 | 390 | 67 | |
| GenBank mRNA | BC134670.1 | - | 1 - 2 | 390 | 67 | |
More Information
Set Membership: |
|

MEDAG
Genomic Map