Assay Details
| Gene Symbol: LRRC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083678.1 | NP_001077147.1 | 2 - 3 | 494 | 65 | |
| GenBank mRNA | BC134596.1 | - | 2 - 3 | 494 | 65 | |
More Information
Set Membership: |
|

LRRC24
Genomic Map