Assay Details
| Gene Symbol: PDGFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083706.1 | NP_001077175.1 | 4 - 5 | 690 | 68 | |
| GenBank mRNA | BC134505.1 | - | 4 - 5 | 690 | 68 | |
More Information
Set Membership: |
|

PDGFD
Genomic Map