Assay Details
| Gene Symbol: RILPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083718.1 | NP_001077187.1 | 2 - 3 | 546 | 92 | |
| GenBank mRNA | BC134625.1 | - | 2 - 3 | 546 | 92 | |
More Information
Set Membership: |
|

RILPL2
Genomic Map