Assay Details
| Gene Symbol: NHLRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083723.2 | NP_001077192.1 | 2 - 3 | 389 | 76 | |
| GenBank mRNA | BC134430.1 | - | 2 - 3 | 390 | 76 | |
More Information
Set Membership: |
|

NHLRC2
Genomic Map