Assay Details
| Gene Symbol: SH3RF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083753.2 | NP_001077222.1 | 2 - 3 | 773 | 75 | |
| GenBank mRNA | BC134725.1 | - | 2 - 3 | 774 | 75 | |
More Information
Set Membership: |
|

SH3RF2
Genomic Map