Assay Details
| Gene Symbol: SLC34A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103223.1 | NP_001096693.1 | 10 - 11 | 1252 | 61 | |
| GenBank mRNA | BC151503.1 | - | 10 - 11 | 1252 | 61 | |
More Information
Set Membership: |
|

SLC34A1
Genomic Map