Assay Details
| Gene Symbol: BTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173896.2 | NP_776321.2 | 2 - 3 | 219 | 69 | |
| GenBank mRNA | AF140597.1 | - | 2 - 3 | 165 | 69 | |
| BC133541.1 | - | 2 - 3 | 219 | 69 | ||
More Information
Set Membership: |
|

BTC
Genomic Map