Assay Details
| Gene Symbol: LIMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040512.2 | NP_001035602.2 | 3 - 4 | 343 | 66 | |
| GenBank mRNA | BC116116.1 | - | 3 - 4 | 343 | 66 | |
| BT021808.1 | - | 1 - 2 | 613 | 66 | ||
More Information
Set Membership: |
|

LIMD2
Genomic Map