Assay Details
| Gene Symbol: TMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075385.2 | NP_001068853.1 | 4 - 5 | 460 | 75 | |
| GenBank mRNA | BC133591.1 | - | 4 - 5 | 466 | 75 | |
| BT030539.1 | - | 4 - 5 | 460 | 75 | ||
More Information
Set Membership: |
|

TMX1
Genomic Map