Assay Details
| Gene Symbol: HSDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098871.1 | NP_001092341.1 | 3 - 4 | 365 | 71 | |
| GenBank mRNA | BC142092.1 | - | 3 - 4 | 365 | 71 | |
More Information
Set Membership: |
|

HSDL1
Genomic Map