Assay Details
| Gene Symbol: AMDHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098872.2 | NP_001092342.1 | 3 - 4 | 331 | 64 | |
| GenBank mRNA | BC142251.1 | - | 3 - 4 | 332 | 64 | |
More Information
Set Membership: |
|

AMDHD1
Genomic Map