Assay Details
| Gene Symbol: ACOT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098941.2 | NP_001092411.2 | 2 - 3 | 785 | 60 | |
| GenBank mRNA | BC142161.1 | - | 2 - 3 | 785 | 60 | |
More Information
Set Membership: |
|

ACOT4
Genomic Map