Assay Details
| Gene Symbol: URAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099703.2 | NP_001093173.1 | 1 - 2 | 205 | 110 | |
| GenBank mRNA | BC142055.1 | - | 1 - 2 | 206 | 110 | |
More Information
Set Membership: |
|

URAD
Genomic Map