Assay Details
| Gene Symbol: NXT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100353.1 | NP_001093823.1 | 2 - 3 | 252 | 92 | |
| GenBank mRNA | BC149046.1 | - | 2 - 3 | 252 | 92 | |
More Information
Set Membership: |
|

NXT2
Genomic Map