Assay Details
| Gene Symbol: DESI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100384.2 | NP_001093854.1 | 1 - 2 | 60 | 70 | |
| GenBank mRNA | BC149676.1 | - | 1 - 2 | 61 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149676.1, NM_001100384.2 transcripts.
Set Membership: |
|

DESI2
Genomic Map