Assay Details
| Gene Symbol: DESI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100384.2 | NP_001093854.1 | 4 - 5 | 363 | 116 | |
| GenBank mRNA | BC149676.1 | - | 4 - 5 | 364 | 116 | |
More Information
Set Membership: |
|

DESI2
Genomic Map