Assay Details
| Gene Symbol: MFNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101051.1 | NP_001094521.1 | 4 - 5 | 724 | 56 | |
| GenBank mRNA | BC116066.1 | - | 4 - 5 | 734 | 56 | |
| BT030676.1 | - | 4 - 5 | 724 | 56 | ||
More Information
Set Membership: |
|

MFNG
Genomic Map