Assay Details
| Gene Symbol: ZCCHC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101055.2 | NP_001094525.1 | 4 - 5 | 718 | 130 | |
| GenBank mRNA | BC146120.1 | - | 4 - 5 | 720 | 130 | |
More Information
Set Membership: |
|

ZCCHC9
Genomic Map