Assay Details
| Gene Symbol: MAP1LC3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101058.2 | NP_001094528.1 | 1 - 2 | 187 | 136 | |
| GenBank mRNA | BC151421.1 | - | 1 - 2 | 188 | 136 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC151421.1, NM_001101058.2 transcripts.
Set Membership: |
|

MAP1LC3C
Genomic Map