Assay Details
| Gene Symbol: MAP1LC3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101058.2 | NP_001094528.1 | 4 - 5 | 431 | 88 | |
| GenBank mRNA | BC151421.1 | - | 4 - 5 | 432 | 88 | |
More Information
Set Membership: |
|

MAP1LC3C
Genomic Map