Assay Details
| Gene Symbol: NAIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101074.2 | NP_001094544.1 | 1 - 2 | 621 | 55 | |
| GenBank mRNA | BC151557.1 | - | 1 - 2 | 622 | 55 | |
More Information
Set Membership: |
|

NAIF1
Genomic Map