Assay Details
| Gene Symbol: FARSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101107.2 | NP_001094577.1 | 7 - 8 | 888 | 69 | |
| GenBank mRNA | BC151497.1 | - | 7 - 8 | 889 | 69 | |
More Information
Set Membership: |
|

FARSA
Genomic Map