Assay Details
Gene Symbol: ZRANB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101114.1 | NP_001094584.1 | 5 - 6 | 1452 | 72 | |
GenBank mRNA | BC149099.1 | - | 5 - 6 | 1452 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
