Assay Details
| Gene Symbol: AMIGO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101119.1 | NP_001094589.1 | 1 - 2 | 291 | 68 | |
| GenBank mRNA | BC151605.1 | - | 1 - 2 | 291 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC151605.1, NM_001101119.1 transcripts.
Set Membership: |
|

AMIGO1
Genomic Map