Assay Details
| Gene Symbol: NINJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101914.2 | NP_001095384.1 | 2 - 3 | 554 | 69 | |
| GenBank mRNA | BC148125.1 | - | 2 - 3 | 555 | 69 | |
More Information
Set Membership: |
|

NINJ2
Genomic Map