Assay Details
| Gene Symbol: FAM149A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101965.1 | NP_001095435.1 | 1 - 2 | 97 | 66 | |
| GenBank mRNA | BC149139.1 | - | 1 - 2 | 97 | 66 | |
More Information
Set Membership: |
|

FAM149A
Genomic Map