Assay Details
| Gene Symbol: TMEM150B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102025.2 | NP_001095495.1 | 6 - 7 | 539 | 66 | |
| GenBank mRNA | BC151463.1 | - | 6 - 7 | 540 | 66 | |
More Information
Set Membership: |
|

TMEM150B
Genomic Map