Assay Details
| Gene Symbol: KLHL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102043.2 | NP_001095513.1 | 8 - 9 | 1263 | 64 | |
| GenBank mRNA | BC151597.1 | - | 8 - 9 | 1264 | 64 | |
More Information
Set Membership: |
|

KLHL18
Genomic Map