Assay Details
Gene Symbol: ADH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102059.2 | NP_001095529.1 | 5 - 6 | 664 | 63 | |
GenBank mRNA | BC149290.1 | - | 5 - 6 | 665 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
