Assay Details
| Gene Symbol: BORCS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145790.1 | NP_001139262.1 | 3 - 4 | 249 | 66 | |
| NM_001145791.1 | NP_001139263.1 | 3 - 4 | 249 | 66 | ||
| NM_001145792.1 | NP_001139264.1 | 3 - 4 | 249 | 66 | ||
| GenBank mRNA | BC122838.1 | - | 3 - 4 | 250 | 66 | |
| EE218140.1 | - | 3 - 4 | 244 | 66 | ||
| EH137931.1 | - | 3 - 4 | 250 | 66 | ||
| FE007848.1 | - | 3 - 4 | 236 | 66 | ||
| Gene Symbol: LOC100271850-MEF2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027315.1 | 3 - 4 | 249 | 66 | ||
More Information
Set Membership: |
|

BORCS8
Genomic Map