Assay Details
| Gene Symbol: C8G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110076.2 | NP_001103546.1 | 5 - 6 | 881 | 77 | |
| GenBank mRNA | BC105414.1 | - | 5 - 6 | 882 | 77 | |
More Information
Set Membership: |
|

C8G
Genomic Map