Assay Details
| Gene Symbol: MPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113298.2 | NP_001106769.1 | 8 - 9 | 1162 | 63 | |
| GenBank mRNA | BC149472.1 | - | 8 - 9 | 1163 | 63 | |
More Information
Set Membership: |
|

MPO
Genomic Map