Assay Details
| Gene Symbol: CPXM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015642.2 | NP_001015642.2 | 12 - 13 | 1872 | 58 | |
| GenBank mRNA | BC133527.1 | - | 12 - 13 | 1894 | 58 | |
| BE751239.1 | - | 2 - 3 | 403 | 58 | ||
| DN544938.1 | - | 2 - 3 | 487 | 58 | ||
More Information
Set Membership: |
|

CPXM1
Genomic Map