Assay Details
| Gene Symbol: PDGFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017953.2 | NP_001017953.2 | 4 - 5 | 461 | 87 | |
| GenBank mRNA | BC151383.1 | - | 4 - 5 | 461 | 87 | |
| BT021749.1 | - | 4 - 5 | 501 | 87 | ||
More Information
Set Membership: |
|

PDGFB
Genomic Map