Assay Details
| Gene Symbol: OLFML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035385.2 | NP_001030462.2 | 1 - 2 | 530 | 70 | |
| GenBank mRNA | BC109584.1 | - | 1 - 2 | 533 | 70 | |
| BT021753.1 | - | 1 - 2 | 502 | 70 | ||
| EE350823.1 | - | 1 - 2 | 282 | 70 | ||
More Information
Set Membership: |
|

OLFML1
Genomic Map