Assay Details
| Gene Symbol: OLFML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035385.2 | NP_001030462.2 | 2 - 3 | 816 | 95 | |
| GenBank mRNA | BC109584.1 | - | 2 - 3 | 819 | 95 | |
| BT021753.1 | - | 2 - 3 | 788 | 95 | ||
| EE350823.1 | - | 2 - 3 | 568 | 95 | ||
| EE382024.1 | - | 1 - 2 | 122 | 95 | ||
More Information
Set Membership: |
|

OLFML1
Genomic Map