Assay Details
| Gene Symbol: SHCBP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046290.2 | NP_001039755.2 | 5 - 6 | 1114 | 77 | |
| GenBank mRNA | BC112556.1 | - | 5 - 6 | 1115 | 77 | |
More Information
Set Membership: |
|

SHCBP1L
Genomic Map